Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium iodide 20% (w/v) in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International
Synonyms: Dipotassium diiodideKali iodide

BDHVBDH7356-1
BDH7356-1CS 2321.8 USD InStock
BDH7356-1
Potassium iodide 20% (w/v) in aqueous solution, VWR Chemicals BDH®
Potassium iodide
Made with deionized water.


Formula: Kl
MW: 166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4
Order Now
Specification Test Results
Appearance/color Clear liquid
Shelf-life 1 year

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty