Synonyms:
Hydrogen sulfateOil of vitriolVitriol brown oil
BDHVBDH7370-4
BDHVBDH7370-1
BDH7370-1CS
723.11
USD
InStock
BDH7370-1
BDH7370-4
Sulfuric acid 20% (v/v) (1+4), VWR Chemicals BDH®
Sulfuric acid
Per liter formula:
200mLs ACS Sulfuric Acid 95-98% W/W
800mLs Deionized Water