Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfuric acid 50% (v/v) (1+1), VWR Chemicals BDH®

Supplier: VWR International
Danger
Synonyms: Hydrogen sulfateOil of vitriolVitriol brown oil

BDHVBDH7371-4 BDHVBDH7371-2
BDH7371-2CS 634.45 USD InStock
BDH7371-2 BDH7371-4
Sulfuric acid 50% (v/v) (1+1), VWR Chemicals BDH®
Sulfuric acid
Per liter formula:
500mLs ACS Sulfuric Acid 95-98% W/W
500mLs Deionized Water


Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: ∼100 °C (1013 hPa)
Density: 1.1…1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II
Order Now
Specification Test Results
Appearance/color Clear liquid
Shelf-life 2 years

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty