Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Citric acid 1 M in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International
Danger
Synonyms: Hydroxytricarballylic acid

BDHVBDH7397-1
BDH7397-1CS 915.73 USD InStock
BDH7397-1
Citric acid 1 M in aqueous solution, VWR Chemicals BDH®
Citric acid
Made with deionized water.


Formula: HOC(COOH)(CH₂COOH)₂
MW: 192.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011669
CAS Number: 77-92-9
EINECS: 201-069-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/50/46/725046.jpg
Order Now
Specification Test Results
Appearance/color Clear pale yellow liquid
N.I.S.T. traceable to SRM 84 Yes
Molarity 0.995 - 1.005 M
Shelf-life 1 year

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty