Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 3.0 N in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International
Danger
Synonyms: Caustic sodaLyeSodium hydrateSoda lye

BDHVBDH7472-1
BDH7472-1CS 1117.98 USD InStock
BDH7472-1
Sodium hydroxide 3.0 N in aqueous solution, VWR Chemicals BDH®
Sodium hydroxide
Made with deionized water. Suitable as a titrant.


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 109 °C (1013 hPa)
Density: ≤1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
Appearance/color Clear liquid
N.I.S.T. traceable to SRM 84 Yes
Normality 2.995 - 3.005 N
Shelf-life 2 years

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty