Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EDTA disodium salt 0.05 M in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International
Synonyms: Edetic acid disodium saltDisodium dihydrogen ethylenediaminetetraacetateEthylenediaminetetra-acetic acid disodium saltIdranal IIIKestranal® 2SEthylenedinitrilotetra-acetic acid disodium salt

BDHVBDH7497-4 BDHVBDH7497-1
BDH7497-1CS 946.68 USD InStock
BDH7497-1 BDH7497-4
EDTA disodium salt 0.05 M in aqueous solution, VWR Chemicals BDH®
EDTA disodium salt
Made with deionized water.

Suitable as a titrant


Formula: C₁₀H₁₄N₂Na₂O₈
MW: 336.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070672
CAS Number: 139-33-3
EINECS: 205-358-3
Order Now
Specification Test Results
Appearance/color Clear liquid
N.I.S.T. traceable to SRM 915 Yes
Molarity 0.0495 - 0.0505 M
Shelf-life 1 year

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty