Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfuric acid 2.0 N in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International
Warning
Synonyms: Hydrogen sulfateOil of vitriolVitriol brown oil

BDHVBDH7500-1
BDH7500-1CS 879.47 USD InStock
BDH7500-1
Sulfuric acid 2.0 N in aqueous solution, VWR Chemicals BDH®
Sulfuric acid
Made with high purity acid and deionized water. Suitable as a titrant.


Formula: H₂SO₄
MW: 98.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II
Order Now
Specification Test Results
Appearance/color Clear liquid
N.I.S.T. Traceable to SRM 723 Yes
Normality 1.95 - 2.05 N
Shelf-life 18 months

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty