Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl orange 0.1% (w/v) in aqueous solution indicator, VWR Chemicals BDH®

Supplier: VWR International
Synonyms: 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium saltMethyl orange sodium saltHelianthine4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulfonic acid, sodium salt

BDHVBDH7610-2 BDHVBDH7610-1
BDH7610-2CS 467.68 USD InStock
BDH7610-2 BDH7610-1
Methyl orange 0.1% (w/v) in aqueous solution indicator, VWR Chemicals BDH®
Methyl orange
Made with deionized water.


Formula: C₁₄H₁₄N₃NaO₃S
MW: 327.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007502
CAS Number: 547-58-0
EINECS: 208-925-3
Order Now
Specification Test Results
Appearance/color Clear orange liquid
Shelf-life 18 months

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty