Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonia solution 5.0 N, VWR Chemicals BDH®

Supplier: VWR International
Warning
Synonyms: Ammonium hydroxideAmmonium hydroxide 10% aqueous solutionAmmonia solution 10%

BDHVBDH7615-1
BDH7615-1CS 1228.36 USD InStock
BDH7615-1
Ammonia solution 5.0 N, VWR Chemicals BDH®
Ammonia solution
Suitable as a titrant


Formula: NH₃
MW: 35.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III
Order Now
Specification Test Results
Appearance/color Clear liquid
N.I.S.T. Traceable to SRM 723 Yes
Normality 4.980 - 5.020 N
Shelf-life 2 years

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty