Synonyms:
Edetic acid disodium salt
, Disodium dihydrogen ethylenediaminetetraacetate
, Ethylenediaminetetra-acetic acid disodium salt
, Idranal III
, Kestranal® 2S
, Ethylenedinitrilotetra-acetic acid disodium salt
BDHVBDH7621-1
BDHVBDH7621-4
BDH7621-1CS
611.81
USD
InStock
BDH7621-1
BDH7621-4
EDTA disodium salt 0.0100 M in aqueous solution, VWR Chemicals BDH®
EDTA disodium salt
Suitable as a titrant. Made with deionized water.