Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hydroxide solution 0.1 N in 2-propanol, VWR Chemicals BDH®

Supplier: VWR International
Synonyms: Caustic potashPotassium hydrate

BDHVBDH7630-1
BDH7630-1CS 2075.68 USD InStock
BDH7630-1
Potassium hydroxide solution 0.1 N in 2-propanol, VWR Chemicals BDH®
Potassium hydroxide


Formula: KOH
MW: 56.11 g/mol
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1813
ADR: 8,II
Order Now
Specification Test Results
Appearance/color Clear to turbid liquid
N.I.S.T. Traceable Yes
Normality 0.0995 - 0.1005 N
Shelf-life 1 year

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty