Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromate-Bromide solution 0.1 N, VWR Chemicals BDH®

Supplier: VWR International
Synonyms: Bromide-Bromate

BDHVBDH7862-1
BDH7862-1CS 1278.21 USD InStock
BDH7862-1
Bromate-Bromide solution 0.1 N, VWR Chemicals BDH®
Bromate-Bromide
APHA for phenols.


Order Now
Specification Test Results
Appearance/color Clear liquid
Normality 0.0995 - 0.1005 N

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty