Synonyms:
2-HydroxypropaneIPAIsopropanolIsopropyl alcoholsec-PropanolPropan-2-olPropane, 2-hydroxy-sec-Propyl AlcoholRubbing Alcohol
BDHVBDH7999-10
BDHVBDH7999-4
BDH7999-4CS
291.41
USD
InStock
BDH7999-4
BDH7999-10
2-Propanol ≥70% (v/v) (3+7), VWR Chemicals BDH®
2-Propanol
Per liter formula:
700mLs ACS Isopropyl Alcohol minimum 99.0% W/W
300mLs Deionized Water