Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,2,4-Trimethylpentane ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®

Supplier: VWR International
Danger
Synonyms: 2,2,4-trimethylpentaneIsobutyltrimethylethaneIsobutyltrimethylmethane

BDH83630.400
BDH83630.400CS 1123.45 USD InStock
BDH83630.400
2,2,4-Trimethylpentane ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
2,2,4-Trimethylpentane


Formula: (CH₃)₂CHCH₂C(CH₃)₃
MW: 114.23 g/mol
Boiling Pt: 99.2 °C (1013 hPa)
Melting Pt: –107 °C
Density: 0.692 g/cm³ (20 °C)
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00008943
CAS Number: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3,II
Merck Index: 13,05212
https://us-legacy.vwr.com/stibo/web/std.lang.all/00/81/740081.jpg
Order Now
Specification Test Results
Assay (by GC-FID) Min 99.5 %
Water (by KF coulometry) Max. 0.0100 %
Nonvolatile Residue Max. 0.0005 %
Acidity Max. 0.0005 meq/g
Alkalinity Max. 0.0002 meq/g
Transmittance (220 nm) Min. 70 %
Transmittance (235 nm) Min. 80 %
Transmittance (255 nm) Min. 98 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty