Synonyms:
CyanomethaneEthanenitrileEthanoic acid nitrileMethane, cyano-Methyl cyanideMethanecarbonitrileACN
Filtered through a 0.2 µm filter, packaged under nitrogen.
BDHVBDH83639.201
BDHVBDH83639.050
BDHVBDH83639.202
BDHVBDH83639.402
BDHVBDH83639.400
BDHVBDH83639.100E
BDH83639.050EA
3828.65
USD
InStock
BDH83639.050
BDH83639.201
BDH83639.202
BDH83639.400
BDH83639.100E
BDH83639.402
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.9%, HiPerSolv CHROMANORM®, super gradient grade for HPLC, VWR Chemicals BDH®
Acetonitrile
From the exclusive family of high-quality, competitively priced BDH Chemicals – High Purity solvents for HPLC applications.
- Designed to meet demanding HPLC and UV/Vis analytical and quality control requirements
- Filtered down to 0.2 micron level and bottled under nitrogen
- Feature tamper-evident caps for added peace of mind
- Carry Globally Harmonized System (GHS) labeling to help with compliance
- Feature environmentally friendly corrugated/pulp case inserts that are fully recyclable
Ordering information: To purchase these solvents in stainless steel returnable drums as part of the VWR ReCAP program, please contact your local VWR sales representative. A separate contract for VWR ReCAP products is required.