Would you like to log in again?
VWR® Volumetric Reagents for Water Determination by the Karl Fischer Method
Supplier: Avantor
VWR®
0009124801BD BDHVBDH85451.320 0008264802BD BDHVBDH85463.320 0009094801BD 0009114801BD 0009174801BD 0008674801BD 0009104801BD BDHVBDH85452.320 0009084801BD BDHVBDH85469.290 BDHVBDH85452.290 BDHVBDH85468.290 BDHVBDH85463.290 0009214801BD 0009084802BD 0009194801BD 0009114802BD 0008264801BD 0009184801BD
BDH85469.290CS 1388.53 USD InStock
BDH85469.290 BDH85462.290 BDH85454.320 BDH85456.290 BDH85452.320 BDH85458.290 BDH85459.290 BDH85460.290 BDH85463.320 BDH85453.290 BDH85457.320 BDH85464.290 BDH85453.320 BDH85468.290 BDH85463.290 BDH85457.290 BDH85455.290 BDH85451.320 BDH85454.290 BDH85461.290 BDH85452.290
VWR® Volumetric Reagents for Water Determination by the Karl Fischer Method
Karl Fischer Titrants and Reagents
These reagents are ideal for water determination including the analysis of aldehydes and ketones.
  • Fast and stable endpoints for accurate results time and time again
  • Low toxicity and pyridine-free
  • Provide consistent results
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty