Would you like to log in again?
VWR® Coulometric Reagents for Water Determination by the Karl Fischer Method
Supplier: Avantor
VWR®
0008654839BD 0008634839BD 00092448G5BD 0008644801BD 00092648G5BD 00092548G5BD 00092748G5BD 00092848G5BD 00086248G5BD 00087848G5BD
BDH85479.130CS 2605.59 USD InStock
BDH85479.130 BDH85478.260 BDH85475.260 BDH85472.260 BDH85470.290 BDH85473.260 BDH85474.260 BDH85476.260 BDH85471.130 BDH85477.260
VWR® Coulometric Reagents for Water Determination by the Karl Fischer Method
Karl Fischer Titrants and Reagents
Both anolyte and catholyte solutions are with improved formulation to increase the speed and accuracy of titration when determining water content at the microgram level.
  • Reach endpoint quickly and consistently
  • Long shelf life
  • Stable endpoint
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty