Would you like to log in again?
TITRINORM® pH Buffers, for Karl Fischer Titrations
Supplier: Avantor
0008514801BD
BDH85498.290CS 734.4 USD InStock
BDH85498.290
TITRINORM® pH Buffers, for Karl Fischer Titrations
Karl Fischer Titrants and Reagents
The volumetric and coulometric reagents are buffered systems, stabilized around the ideal value between 5 to 7 ensuring a rapid and stoichiometric course of reaction. Buffering systems which neutralize and stabilize the pH during the reaction are suitable for this purpose.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty