Synonyms:
CyanomethaneEthanenitrileEthanoic acid nitrileMethane, cyano-Methyl cyanideMethanecarbonitrileACN
BDHVBDH85501.201
BDHVBDH85501.400
BDHVBDH85501.050
BDHVBDH85501.202
BDHVBDH85501.100E
BDH85501.100ECS
581.26
USD
InStock
BDH85501.100E
BDH85501.050
BDH85501.400
BDH85501.202
BDH85501.201
Acetonitrile, anhydrous ≥99.9% for DNA synthesis
Acetonitrile
From the exclusive family of high-quality, competitively priced BDH Chemicals – High Purity solvents for HPLC applications.
Solvents are pumped through 0.2 micron filters on filling and packaged under Nitrogen.
Packaging: All bottles have tamper evident caps for added peace of mind and are packed in environmentally –friendly corrugated/pulp case inserts that are fully recyclable.