Would you like to log in again?
Platinum, Single Element ICP and ICP/MS Certified Reference Standards, Enhanced Packaging, ARISTAR®, VWR Chemicals BDH®
Supplier: Avantor
VWR Chemicals BDH® ARISTAR®
CABDH89800-114 CABDH89800-118 BDH89800-424 CABDH89800-426 BDH82026-036 MSPT-100PPM-125ML BDH89800-118 CABDH82026-036 BDH89800-426 BDH89800-254 CABDH82026-038 CABDH89800-116 CABDH89800-112 CABDH89800-424 BDH89800-112 BDH89800-114 BDH89800-226 BDH89800-116 BDH82026-038 MSPT-10PPM-125ML
BDH82026-036EA 125 USD InStock
CABDH89800-118 CABDH89800-426 CABDH89800-116 CABDH89800-424 CABDH89800-226 CABDH89800-114 BDH82026-036 CABDH89800-112 CABDH89800-254 BDH82026-038 BDH89800-118 BDH89800-426 BDH89800-424 BDH89800-226 BDH89800-116 BDH89800-114 CABDH82026-038 BDH89800-112 CABDH82026-036 BDH89800-254
Platinum, Single Element ICP and ICP/MS Certified Reference Standards, Enhanced Packaging, ARISTAR®, VWR Chemicals BDH®
Standards Cationic Standards Platinum Standards
Used with ICP-MS, ICP-OES, DCP, GFAA, and AA
  • Long shelf life
  • NIST traceable
  • Precise and accurate
These ICP and ICP-MS CRMs are engineered to be stable, compatible, traceable to NIST SRMs, and are manufactured and tested under ISO Guide 34 & ISO 17025 guidelines. Certified values are based on two independent methods, uncertainties are based on detailed error budgets, and trace metallic impurities (TMI) are determined by ICP and ICP-MS for single element CRMs
Each single element ICP standard ships with a Certificate of Analysis (C of A) and Safety Data Sheet (SDS). Specific lot C of A’s can also be found online.
Enhanced packaging extends the shelf life to 4 years by sealing the product in an external aluminized bag over the regular poly bottle.
This specially designed outer bag prevents transpiration (the loss of water, or solvent, through the container.) Therefore, most of these standards have a 4 year shelf life until the outer bag is opened. After opening the outer bag, the customer will mark the inner bottle with a one-year shelf life from that date.
Single element Certified Reference Materials (CRMs) are available in the BDH range for most of the periodic table, in concentrations ranging from 10, 100, 1,000, and 10,000 µg/mL
Please note: 1000 ppm = 1000 µg/mL
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty