Would you like to log in again?
VWR® Yttrium, Single Element ICP and ICP/MS Certified Reference Standards, Enhanced Packaging
Supplier: Avantor
CABDH82026-090 MSY-100PPM-125ML BDH89800-536 CABDH89800-536 BDH89800-202 CABDH89800-202 BDH82026-090 BDH82026-268 MSY-10PPM-125ML BDH82026-266 CABDH89800-538 BDH89800-538
BDH89800-538EA 303 USD InStock
BDH89800-538 BDH89800-536 CABDH89800-538 CABDH89800-536 CABDH82026-266 BDH89800-202 CABDH89800-202 BDH82026-268 CABDH82026-268 BDH82026-266 BDH82026-090 CABDH82026-090
VWR® Yttrium, Single Element ICP and ICP/MS Certified Reference Standards, Enhanced Packaging
Standards Cationic Standards Yttrium Standards
Used with ICP-MS, ICP-OES, DCP, GFAA, and AA
  • Long shelf life
  • NIST traceable
  • Precise and accurate
These ICP and ICP-MS CRMs are engineered to be stable, compatible, traceable to NIST SRMs, and are manufactured and tested under ISO Guide 34 & ISO 17025 guidelines. Certified values are based on two independent methods, uncertainties are based on detailed error budgets, and trace metallic impurities (TMI) are determined by ICP and ICP-MS for single element CRMs
Each single element ICP standard ships with a Certificate of Analysis (C of A) and Safety Data Sheet (SDS). Specific lot C of A’s can also be found online.
Enhanced packaging extends the shelf life to 4 years by sealing the product in an external aluminized bag over the regular poly bottle.
This specially designed outer bag prevents transpiration (the loss of water, or solvent, through the container.) Therefore, most of these standards have a 4 year shelf life until the outer bag is opened. After opening the outer bag, the customer will mark the inner bottle with a one-year shelf life from that date.
Single element Certified Reference Materials (CRMs) are available in the BDH range for most of the periodic table, in concentrations ranging from 10, 100, 1,000, and 10,000 µg/mL
Please note: 1000 ppm = 1000 µg/mL
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty