Synonyms:
Sodium bicarbonate
BDHVBDH9280-500G
BDHVBDH9280-12KG
BDHVBDH9280-2.5KG
BDH9280-2.5KGCS
349.22
USD
InStock
BDH9280-2.5KG
BDH9280-12KG
BDH9280-500G
Sodium hydrogen carbonate 99.7-100.3% ACS, VWR Chemicals BDH®
Sodium hydrogen carbonate
Meets reagent specifications for testing USP/NF monographs.
White solid