Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trisodium phosphate dodecahydrate 98.0-102.0% ACS, VWR Chemicals BDH®

Supplier: VWR International

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Warning
Synonyms: Sodium Phosphate tribasic 12-HydrateSodium phosphate tribasic dodecahydrateTrisodium phosphateSodium phosphate dodecahydrate

BDHVBDH9300-500G
BDH9300-500GCS 948.14 USD InStock
BDH9300-500G
Trisodium phosphate dodecahydrate 98.0-102.0% ACS, VWR Chemicals BDH®
tri-Sodium phosphate dodecahydrate
Meets reagent specifications for testing USP/NF monographs.

Colorless or white solid


Formula: Na₃PO₄·12H₂O
MW: 380.12 g/mol
Melting Pt: ∼75 °C
Density: 1.62 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149198
CAS Number: 10101-89-0
EINECS: 231-509-8
ADR: 8,III
REACH: 01-2119489800-32-xxxx
Merck Index: 12,08808
Order Now
Specification Test Results
Chloride = 0.001 %
Excess Alkali = 2.5 %
Heavy Metals = 0.001 %
Insolubles = 0.01 %
Iron = 0.001 %
Purity 98.0 - 102.0 %
Sulfate = 0.01 %
Meets reagent specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty