Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium sulfate, anhydrous ≥99.0%, white powder ACS, VWR Chemicals BDH®

Supplier: VWR International
Synonyms: Sulphuric acid disodium saltBisodium sulphateSulfuric acid disodium saltBisodium sulfateDisodium sulphateDisodium sulfate

BDHVBDH9302-2.5KG BDHVBDH9302-12KG BDHVBDH9302-500G
BDH9302-500GCS 663.33 USD InStock
BDH9302-500G BDH9302-12KG BDH9302-2.5KG
Sodium sulfate, anhydrous ≥99.0%, white powder ACS, VWR Chemicals BDH®
Sodium sulphate


Formula: Na₂SO₄
MW: 142.04 g/mol
Boiling Pt: 1429 °C (1013 hPa)
Melting Pt: 888 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003504
CAS Number: 7757-82-6
EINECS: 231-820-9
Merck Index: 12,08829
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/41/16640641.jpg
Order Now
Specification Test Results
Calcium <= 0.01 %
Chloride <= 0.001 %
Heavy Metals (as Pb) <= 0.0005 %
Insolubles <= 0.01 %
Iron <= 0.001 %
Loss on Ignition <= 0.5 %
Magnesium <= 0.005 %
Nitrogen Compounds <= 0.0005 %
pH (5%, Water) @25C 5.2 - 9.2
Phosphate <= 0.001 %
Potassium <= 0.01 %
Purity >= 99.0 %
Meets reagent specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty