Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(+)-Sucrose ACS, VWR Chemicals BDH®

Supplier: VWR International
Synonyms: alpha-D-GlucopyranosideSucroseD-(+)-SaccharoseD(+)-Saccharosebeta-D-Fructofuranosyl alpha-D-glucopyranosideSugar

BDHVBDH9308-2.5KG BDHVBDH9308-500G
BDH9308-2.5KGEA 353.64 USD InStock
BDH9308-2.5KG BDH9308-500G
D-(+)-Sucrose ACS, VWR Chemicals BDH®
D(+)-Sucrose
White solid


Formula: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
Melting Pt: 169…170 °C
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS Number: 57-50-1
EINECS: 200-334-9
Merck Index: 12,09051
https://us-legacy.vwr.com/stibo/web/std.lang.all/95/86/2299586.jpg
Order Now
Specification Test Results
Chloride = 0.005 %
Heavy Metals (as Pb) = 0.0005 %
Insolubles = 0.005 %
Invert Sugar = 0.05 %
Iron = 0.0005 %
Loss on Drying = 0.03 %
Residue after Ignition = 0.01 %
Specific Rotation +66.3 - +66.8 degrees
Sulfate & Sulfite = 0.005 %
Titrable Free Acid = 0.0008 meq/g

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty