Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Acetonitrile ≥99.9% (by GC), CHROMASOLV™, gradient grade suitable for PAH analysis, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Methane, cyano-Methyl cyanideEthanoic acid nitrileMethanecarbonitrileEthanenitrileCyanomethaneACN

00683-4X4L 00683-2.5L
BJ00683-2.5LEA 404.28 USD InStock
BJ00683-2.5L BJ00683-4X4L
Acetonitrile ≥99.9% (by GC), CHROMASOLV™, gradient grade suitable for PAH analysis, Riedel-de Haën™
Acetonitrile
  • Same high quality as CHROMASOLV HPLC
  • Additional testing for HPLC via gradient analysis and UV-VIS spectroscopy
  • Low UV profile by HPLC
  • Low impurity gradient LC baseline

Beilstein Number: 741857


Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 82 °C (1.013 hPa)
Melting Pt: -46 °C
Density: 0.7857 g/cm³ (20 °C)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/10/718910.jpg
Order Now
Specification Test Results
Assay (GC) Min. 99.9 %
Density (D 20/4) 0.765 - 0.794
Refractive index (n 20/D) 1.343 - 1.345
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.02 %
Free acid (as CH₃COOH) Max. 0.005 %
Free alkali (as NH₃) Max. 0.0005 %
Suitability test (HPLC) complying
Absorbance at 230 nm Max. 0.005
Absorbance at 215 nm Max. 0.01
Absorbance at 200 nm Max. 0.04
Fluorescent substances complying
Refractive Index n20/D 1.344 (lit.)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty