Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Acetonitrile ≥99.9%, B&J Brand™, meets UV Specifications for applications requiring low aldehydes & ketones carbonyl-free, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Methane, cyano-Methyl cyanideEthanoic acid nitrileMethanecarbonitrileEthanenitrileCyanomethaneACN

018-4 018-1L 018-4 018-1L
BJ018-1LCS 886.44 USD InStock
BJ018-1L CABJ018-4 BJ018-4 CABJ018-1L
Acetonitrile ≥99.9%, B&J Brand™, meets UV Specifications for applications requiring low aldehydes & ketones carbonyl-free, Burdick & Jackson™
Acetonitrile
Acetonitrile carbonyl-Free, Meets UV Specifications.

  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency


Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 82 °C (1.013 hPa)
Melting Pt: -46 °C
Density: 0.7822 g/cm³ (20 °C)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/10/718910.jpg
Order Now
Specification Test Results
Carbonyl Compounds (as HCHO) 10 ppb max.
Water by Karl Fischer Titration 0.01 % max.
GC Analysis 99.9 % min.
HPLC Gradient @ 215 nm 0.005 AU max.
HPLC Gradient @ 254 nm 0.001 AU max.
Refractive Index (20ºC) 1.3434 - 1.3446
Residue 1 mg/L max.
UV Absorbance @ 200 nm 0.05 AU max.
UV Absorbance @ 210 nm 0.04 AU max.
UV Cutoff 190 nm max.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty