Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide monohydrate ≥99.9995% (metals basis), TraceSELECT™ for trace analysis, Fluka™

Supplier: Honeywell Research Chemicals
Danger

01968-100G 01968-25G
BJ01968-100GEA 279.01 USD InStock
BJ01968-100G BJ01968-25G
Sodium hydroxide monohydrate ≥99.9995% (metals basis), TraceSELECT™ for trace analysis, Fluka™
Sodium hydroxide monohydrate
TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions


Formula: HNaO·1H₂O
Boiling Pt: 1.390 °C (1.013 hPa. Anhydrous substance)
Melting Pt: 319 °C (anhydr.)
Storage Temperature: Ambient
MDL Number: MFCD00149229
CAS Number: 12179-02-1
EINECS: 215-185-5
UN: 1823
ADR: 8,II
Order Now
Specification Test Results
Aluminium (Al) Max. 0.05 ppm
Arsenic (As) Max. 0.01 ppm
Barium (Ba) Max. 0.5 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.01 ppm
Cerium (Ce) Max. 0.01 ppm
Cobalt (Co) Max. 0.01 ppm
Chromium (Cr) Max. 0.01 ppm
Copper (Cu) Max. 0.01 ppm
Europium (Eu) Max. 0.01 ppm
Iron (Fe) Max. 0.05 ppm
Mercury (Hg) Max. 1 ppb
Iridium (Ir) Max. 0.5 ppm
Potassium (K) Max. 2 ppm
Lanthanum (La) Max. 0.01 ppm
Lithium (Li) Max. 0.2 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.01 ppm
Nickel (Ni) Max. 0.01 ppm
Lead (Pb) Max. 0.01 ppm
Palladium (Pd) Max. 0.5 ppm
Platinum (Pt) Max. 0.5 ppm
Rhodium (Rh) Max. 0.5 ppm
Ruthenium (Ru) Max. 0.5 ppm
Antimony (Sb) Max. 0.05 ppm
Scandium (Sc) Max. 0.01 ppm
Selenium (Se) Max. 0.01 ppm
Samarium (Sm) Max. 0.01 ppm
Strontium (Sr) Max. 0.5 ppm
Thallium (Tl) Max. 0.01 ppm
Vanadium (V) Max. 0.1 ppm
Yttrium (Y) Max. 0.01 ppm
Ytterbium (Yb) Max. 0.01 ppm
Zinc (Zn) Max. 0.05 ppm
Silicate (as SiO₂) Max. 5 ppm
Bromide (Br) Max. 0.5 ppm
Chloride (Cl) Max. 1 ppm
Fluoride (F) Max. 0.5 ppm
Nitrite (NO₂) Max. 0.5 ppm
Nitrate (NO₃) Max. 1 ppm
Phosphate (PO₄) Max. 0.05 ppm
Sulfate (SO₄) Max. 2.5 ppm
Total N Max. 5 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty