Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2',7'-Dichlorofluorescein solution indicator, ready-to-use, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: 2',7'-Dichloro-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one

02591-500ML
BJ02591-500MLEA 249.64 USD InStock
BJ02591-500ML
2',7'-Dichlorofluorescein solution indicator, ready-to-use, Fluka™
2',7'-Dichlorofluorescein


Formula: C₂₀H₁₀Cl₂O₅
MW: 401.20 g/mol
Boiling Pt: 81-83 °C
Melting Pt: -90 °C (main component)
Density: 0.79 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00005047
CAS Number: 76-54-0
UN: 1219
ADR: 3,II
Order Now
Specification Test Results
Appearance of the substance complying
Density (D 20/4) 0.775 - 0.804
Absorption maximum 505 - 519 nm
Absorbtion (1%/1cm) Max. 5.0
non-volatile matter Max. 0.001 %
water (Karl Fischer) Max. 0.1 %
titrable acid Max. 0.001 meq/g
aluminium (Al) Max. 0.5 ppm
boron (B) Max. 0.02 ppm
barium (Ba) Max. 0.1 ppm
bismuth (Bi) Max. 0.1 ppm
calcium (Ca) Max. 0.5 ppm
cadmium (Cd) Max. 0.05 ppm
cobalt (Co) Max. 0.02 ppm
chromium (Cr) Max. 0.02 ppm
copper (Cu) Max. 0.02 ppm
iron (Fe) Max. 0.1 ppm
potassium (K) Max. 0.5 ppm
lithium (Li) Max. 0.1 ppm
magnesium (Mg) Max. 0.1 ppm
manganese (Mn) Max. 0.02 ppm
molybdenum (Mo) Max. 0.1 ppm
sodium (Na) Max. 0.5 ppm
nickel (Ni) Max. 0.02 ppm
lead (Pb) Max. 0.1 ppm
tin (Sn) Max. 0.1 ppm
strontium (Sr) Max. 0.1 ppm
zinc (Zn) Max. 0.1 ppm
carbonyl compounds (as HCHO) Max. 100 ppm
APHA Max. 20

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty