Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Iodine 99.8-100.5%, Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™

Supplier: Honeywell Research Chemicals
Danger

03002-6X1KG 03002-500G
BJ03002-500GEA 390.62 USD InStock
BJ03002-500G BJ03002-1KG
Iodine 99.8-100.5%, Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Iodine


Formula: I₂
MW: 253.81 g/mol
Melting Pt: 113.5 °C
Density: 4.930 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2
EINECS: 231-442-4
UN: 3495
ADR: 8,III
Order Now
Specification Test Results
Vapor Density 9 (vs air)
Assay 99.8 - 100.5 %
Non-volatile matter Max. 0.03 %
Bromide, chloride (as Cl) Max. 0.01 %
Residual solvents complying
Vapor Pressure 0.31 mmHg ( 25 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty