Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Dimethylacetamide ≥99.5%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™

Supplier: Honeywell Research Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Acetic acid dimethylamide

071-4 NP071-20 NP071-20 071-1L CS071AD-213 071-4
BJNP071-20EA 3251.73 USD InStock
CABJNP071-20 BJNP071-20 BJ071-1L BJ071-4 CABJ071-4 TXL071AD213DSM
N,N-Dimethylacetamide ≥99.5%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
N,N-Dimethylacetamide
For HPLC, liquid and gas chromatography, pesticide residue analysis, spectophotometry, organic synthesis and combinatorial chemistry.

  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency


Formula: CH₃CON(CH₃)₂
MW: 87.12 g/mol
Boiling Pt: 166 °C
Melting Pt: -20 °C
Density: 0.937 g/cm³ (25 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00008686
CAS Number: 127-19-5
EINECS: 204-826-4
ADR: 6.1,III
Merck Index: 13,03254
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/13/2298913.jpg
Order Now
Specification Test Results
GC Analysis 99.5 % min.
GC Impurities Eluting Before Solvent 0.5 mg/L max.
UV Absorbance @ 275 nm 0.3 AU max.
UV Absorbance @ 300 nm 0.08 AU max.
UV Absorbance @ 350 nm 0.005 AU max.
UV Absorbance @ 400 nm 0.005 AU max.
UV Cutoff 268 nm max.
Water by Karl Fischer Titration 0.03 % max.

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty