Synonyms:
Muriatic acid
08256-2.5L
08256-500ML
08256-100ML
08256-1L
BJ08256-500MLEA
104.2
USD
InStock
BJ08256-500ML
BJ08256-100ML
BJ08256-1L
BJ08256-2.5L
Hydrochloric acid ≥30%, TraceSELECT™ for trace analysis, Fluka™
Hydrochloric acid
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions