Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl acetate ≥99.9%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: EtOAcAcetidinEthyl ethanoateVinegar naphthaEthyl acetic esterAcetic acid ethyl esterAcetic etherAcetoxyethane

100-4 NP100-20 100-4 NP100-20 100-1L
BJNP100-20EA 1128.96 USD InStock
CABJ100-4 BJNP100-20 BJ100-4 BJ100-1L CABJNP100-20
Ethyl acetate ≥99.9%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
Ethyl acetate
For HPLC, liquid and gas chromatography, pesticide residue analysis, spectophotometry, organic synthesis and combinatorial chemistry.

  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency


Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77 °C
Melting Pt: -84 °C
Density: 0.9006 g/cm³ (20 °C)
Flash Pt: -4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://us-legacy.vwr.com/stibo/web/std.lang.all/20/72/722072.jpg
Order Now
Specification Test Results
Electron Capture GC 10 ng/L max.
Water by Karl Fischer Titration 0.03 % max.
GC Analysis 99.9 % min.
Refractive Index (20ºC) 1.3719 - 1.3727
Residue 1 mg/L max.
UV Absorbance @ 275 nm 0.05 AU max.
UV Absorbance @ 300 nm 0.03 AU max.
UV Absorbance @ 325 nm 0.005 AU max.
UV Absorbance @ 400 nm 0.005 AU max.
UV Cutoff 256 nm max.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty