Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver nitrate 99.8-100.5%, meets analytical specification of BP, Ph. Eur., USP, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Silver mononitrateNitric acid silver saltLunar caustic

10220-100G 10220-500G 10220-1KG 10220-50G
BJ10220-1KGEA 2644.43 USD InStock
BJ10220-1KG BJ10220-100G BJ10220-50G BJ10220-500G
Silver nitrate 99.8-100.5%, meets analytical specification of BP, Ph. Eur., USP, Fluka™
Silver nitrate


Formula: AgNO₃
MW: 169.87 g/mol
Melting Pt: 212 °C
Density: 4.350 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 1493
ADR: 5.1,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/45/16640645.jpg
Order Now
Specification Test Results
Assay 99.8 - 100.5 %
Acid. or alk. react. impurities complying
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 0.0005 %
Lead (Pb) Max. 0.002 %
Substances not precipitated by HCl (as SO₄) Max. 0.03 %
Nitrite (NO₂) Max. 0.1 %
Sulfate (SO₄) Max. 0.01 %
Al, Pb, Bi and Cu ions complying
Appearance of the solution complying
Residual solvents complying
Vapor Density 5.8 (vs air)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty