Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Diethyl ether ≥99.9% (excluding ethanol) stabilized, B&J Brand™ for HPLC, for pesticide residue analysis, for spectrophotometry, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Ether2,2'-OxybisethaneEthoxyethaneEthyl Ether2,2'-OxydiethaneEthyl ether

Stabilization: Stabilised with ethanol 2%

106-1L 106-1L 106-4 106-4
BJ106-1LCS 1155.09 USD InStock
CABJ106-4 CABJ106-1L BJ106-1L BJ106-4
Diethyl ether ≥99.9% (excluding ethanol) stabilized, B&J Brand™ for HPLC, for pesticide residue analysis, for spectrophotometry, Burdick & Jackson™
Diethyl ether
For HPLC, liquid and gas chromatography, pesticide residue analysis, spectophotometry, organic synthesis and combinatorial chemistry.

  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency


Formula: (CH₃CH₂)₂O
MW: 74.12 g/mol
Boiling Pt: 35 °C
Melting Pt: -117.4 °C
Density: 0.713 g/cm³ (20 °C)
Flash Pt: -45 °C
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://us-legacy.vwr.com/stibo/web/std.lang.all/02/58/730258.jpg
Order Now
Specification Test Results
Electron Capture GC 10 ng/L max.
Water by Karl Fischer Titration 0.03 % max.
GC Analysis (excluding ethanol) 99.9 % min.
Peroxides (as H₂O₂) at time of packaging 1 ppm max.
Refractive Index (20ºC) 1.3523 - 1.3533
Residue 1 mg/L max.
UV Absorbance @ 250 nm 0.08 AU max.
UV Absorbance @ 275 nm 0.01 AU max.
UV Absorbance @ 300 nm 0.005 AU max.
UV Cutoff 215 nm max.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty