Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Immersion Oil, Honeywell

Supplier: Honeywell Burdick & Jackson

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

10976-20ML
BJ10976-20MLEA 240.57 USD InStock
BJ10976-20ML
Immersion Oil, Honeywell
Stains Microscopy and staining reagents
UV-transparent, fluorescence-free immersion oil.

Specifications
Density (D 20/4): 1.075 - 1.104
Refractive index (n 20/D): 1.516 - 1.518
Fluorescent substances: complying

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty