Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iron(II) sulfate heptahydrate 99.5-104.5% (by manganometry), Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, Fluka™

Supplier: Honeywell Research Chemicals
Warning
Synonyms: Ferrous sulphate heptahydrateIron vitriolFerrous sulfate heptahydrate

12354-4X5KG 12354-6X1KG 12354-500G
BJ12354-5KGCS 1698.48 USD InStock
BJ12354-5KG BJ12354-1KG BJ12354-500G
Iron(II) sulfate heptahydrate 99.5-104.5% (by manganometry), Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, Fluka™
Iron (II) sulphate heptahydrate


Formula: FeSO₄·7H₂O
MW: 278.02 g/mol
Melting Pt: 64 °C
Density: 1.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149719
CAS Number: 7782-63-0
EINECS: 231-753-5
Order Now
Specification Test Results
Assay (manganometric) 99.5 - 104.5 %
pH (5 %, 20°C) 3.0 - 4.0
Arsenic (As) Max. 2 ppm
Calcium (Ca) Max. 0.02 %
Chromium (Cr) Max. 50 ppm
Copper (Cu) Max. 30 ppm
Iron(III) Max. 0.2 %
Mercury (Hg) Max. 1 ppm
Potassium (K) Max. 0.002 %
Magnesium (Mg) Max. 0.02 %
Manganese (Mn) Max. 0.1 %
Sodium (Na) Max. 0.01 %
Nickel (Ni) Max. 50 ppm
Lead (Pb) Max. 2 ppm
Zinc (Zn) Max. 20 ppm
Heavy metals (as Pb) Max. 0.005 %
Chloride (Cl) Max. 100 ppm
Residual solvents complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty