Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium carbonate decahydrate 99.5-100.5% (dried basis), Puriss., meets analytical specification of BP, FCC, Ph. Eur., E500, Fluka™

Supplier: Honeywell Research Chemicals
Warning

13414-6X1KG 13414-2.5KG
BJ13414-1KGCS 324.96 USD InStock
BJ13414-1KG BJ13414-2.5KG
Sodium carbonate decahydrate 99.5-100.5% (dried basis), Puriss., meets analytical specification of BP, FCC, Ph. Eur., E500, Fluka™
Sodium carbonate decahydrate


Formula: Na₂CO₃·10H₂O
MW: 286.14 g/mol
Melting Pt: 32 °C
Density: 1.460 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149178
CAS Number: 6132-02-1
EINECS: 207-838-8
Order Now
Specification Test Results
Assay (excluding dried substance) 99.5 - 100.5 %
Assay of Na₂CO₃ 36.7 - 40.0 %
Loss on drying 55.0 - 65.0 %
Arsenic (As) Max. 2 ppm
Copper (Cu) Max. 0.001 %
Iron (Fe) Max. 5 ppm
Mercury (Hg) Max. 1 ppm
Potassium (K) Max. 0.02 %
Lead (Pb) Max. 2 ppm
Zinc (Zn) Max. 0.001 %
Heavy metals (as Pb) Max. 10 ppm
Chloride (Cl) Max. 30 ppm
Sulfate (SO₄) Max. 100 ppm
Appearance of the solution complying
Alkali hydroxide, -bicarbonates complying
Residual solvents complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty