Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Pentane ≥95.0% (by GC), Laboratory Reagent

Supplier: Honeywell Research Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Pentane

15607-5L 15607-4X5L
BJ15607-5LEA 221.49 USD InStock
BJ15607-5L BJ15607-4X5L
n-Pentane ≥95.0% (by GC), Laboratory Reagent
n-Pentane
  • ≥95% for general laboratory use.

Beilstein Number: 969132
RTECS Number: RZ9450000
UNSPSC Code: 12191502


Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 36-37 °C
Melting Pt: -131 °C
Density: 0.631 g/cm³ (15 °C)
Flash Pt: <-35 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/81/67/738167.jpg
Order Now
Specification Test Results
Assay (GC) Min. 95.0 %
Water (Karl Fischer) Max. 0.02 %
Non-volatile matter Max. 0.002 %
Autoignition Temperature 500 °F
Refractive Index n20/D 1.358 (lit.)
Vapor Density 2.48 (vs air)

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty