Synonyms:
1,4-epoxy-ButaneButane alpha,delta-oxideCyclotetramethylene oxideOxacyclopentaneOxolaneTetramethylene oxideTHF
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 250 ppm
178810-1L
178810-2.5L
178810-20L
178810-10L
BJ178810-2.5LEA
144.17
USD
InStock
BJ178810-2.5L
BJ178810-1L
BJ178810-10L
BJ178810-20L
Tetrahydrofuran ≥99.0% contains inhibitor, Reagent Grade
Tetrahydrofuran