Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Carbon disulfide ≥99.9% (by GC) ACS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

180173-500ML 180173-1L
BJ180173-500MLEA 184.96 USD InStock
BJ180173-500ML BJ180173-1L
Carbon disulfide ≥99.9% (by GC) ACS, Riedel-de Haën™
Carbon disulphide
ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications


Formula: CS₂
MW: 76.14 g/mol
Boiling Pt: 46 °C (1.013 hPa)
Melting Pt: -76 °C
Density: 1.264 g/cm³ (20 °C)
Flash Pt: -30 °C
Storage Temperature: Ambient
MDL Number: MFCD00011321
CAS Number: 75-15-0
EINECS: 200-843-6
UN: 1131
ADR: 3,I
https://us-legacy.vwr.com/stibo/web/std.lang.all/53/94/785394.jpg
Order Now
Specification Test Results
Identity (IR) complying
Assay (GC) Min. 99.90 %
Non-volatile matter Max. 0.002 %
Water (Karl Fischer) Max. 0.05 %
Sulfur dioxide (SO₂) complying
Hydrogen sulfide (H₂S) complying
APHA Max. 10
Autoignition Temperature 212 °F
Refractive Index n20/D 1.627 (lit.)
Explosion Limit 50 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty