Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium oxide activated basic Brockmann Grade I ~150 mesh, Fluka™

Supplier: Honeywell Research Chemicals

199443-5KG 199443-1KG 199443-20KG
BJ199443-1KGEA 150.9 USD InStock
BJ199443-1KG BJ199443-5KG BJ199443-20KG
Aluminium oxide activated basic Brockmann Grade I ~150 mesh, Fluka™
Aluminium oxide activated basic


Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2.980 °C (1.013 hPa)
Melting Pt: 2.072 °C
Density: 3.960 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
Order Now
Specification Test Results
Brockmann Number (Activity) I (val.ref) complying
pH (10 % susp., 20°C) 8.5 - 10.5
Spez. surface (BET) 184.5 - 225.5 m²/g
Fe (as Fe₂O₃) Max. 0.03 %
Na (as Na₂O) Max. 0.4 %
Silicon (as SiO₂) Max. 0.03 %
Chloride (Cl) complying
Grain size 0.04-0.30 mm Min. 95 %
Form powder

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty