Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

n-Hexane ≥95%, Laboratory Reagent

Supplier: Honeywell Research Chemicals
Danger
Synonyms: HexaneHexanenHexyl hydride

208752-5L 208752-1L 208752-4X2.5L 208752-2.5L 208752-190L 208752-20L 208752-25L 208752-4X5L
BJ208752-5LEA 288.66 USD InStock
BJ208752-5L BJ208752-1L BJ208752-2.5L BJ208752-190L BJ208752-20L BJ208752-25L BJ208752-4X5L BJ2087524X2.5L
n-Hexane ≥95%, Laboratory Reagent
n-Hexane
  • ≥95% for general laboratory use.

Beilstein Number: 1730733
RTECS Number: MN9275000
UNSPSC Code: 12191502


Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 66 - 69 °C (1.013 hPa)
Melting Pt: -95 °C
Density: 0.660-0.668 g/cm³ (20 °C)
Flash Pt: -26 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/70/18/737018.jpg
Order Now
Specification Test Results
Autoignition Temperature 453 °F
IR spectrum complying
Assay (GC) Min. 95.0 %
Density (D 20/20) 0.658 - 0.662
Refractive index (n 20/D) 1.373 - 1.377
Non-volatile matter Max. 0.0015 %
Water (Karl Fischer) Max. 0.015 %
Aromatic substances (as C₆H₆) Max. 0.005 %
Vapor Pressure ~132 mmHg ( 20 °C)
Vapor Pressure 256 mmHg ( 37.7 °C)
Explosion Limit 7.7 %
Vapor Pressure 5.2 psi ( 37.7 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty