Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide ≥97.0%, pellets ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

221465-6X500G
BJ221465-500GCS 440.42 USD InStock
BJ221465-500G
Sodium hydroxide ≥97.0%, pellets ACS, Fluka™
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1.390 °C (1.013 hPa)
Melting Pt: 319 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
Order Now
Specification Test Results
Assay Min. 97.0 %
Assay of Na₂CO₃ Max. 1.0 %
Calcium (Ca) Max. 0.005 %
Iron (Fe) Max. 0.001 %
Mercury (Hg) Max. 0.1 ppm
Potassium (K) Max. 0.02 %
Magnesium (Mg) Max. 0.002 %
Nickel (Ni) Max. 0.001 %
Heavy metals (as Ag) Max. 0.002 %
Chloride (Cl) Max. 0.005 %
Phosphate (PO₄) Max. 0.001 %
Sulfate (SO₄) Max. 0.003 %
Total N Max. 0.001 %
Vapor Pressure <18 mmHg ( 20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty