Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

di-Lithium tetraborate ≥99.9% (trace metals basis) PPA

Supplier: Honeywell Research Chemicals
Synonyms: Dilithium tetraborateLithium tetraborate

222534-1KG 222534-250G
BJ222534-1KGEA 616.02 USD InStock
BJ222534-1KG BJ222534-250G
di-Lithium tetraborate ≥99.9% (trace metals basis) PPA
di-Lithium tetraborate


Formula: Li₂B₄O₇
MW: 169.12 g/mol
Melting Pt: 760 - 880 °C
Density: 0.25 g/cm³ (25 °C)
Storage Temperature: Room Temperature
MDL Number: MFCD00011083
CAS Number: 12007-60-2
EINECS: 234-514-3
Order Now
Specification Test Results
Loss on ignition (800 °C, 1 h) Max. 1.00 %
Grain dissipation (medium value) Max. 500 µm
Metal trace analysis (ICP) Max. 1000 ppm
Boron (B) Min. 25.0 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty