Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethyl ether ≥99.5% (by GC) stabilized, Puriss., meets analytical specification of BP, Ph. Eur., Riedel-de Haën™

Supplier: Honeywell Research Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: 2,2'-Oxybisethane2,2'-OxydiethaneEtherEthoxyethaneEthyl ether

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 5 - 8 ppm

24004-1L 24004-135KG 24004-4X5L 24004-25L 24004-5L 24004-6X1L 24004-4X2.5L 24004-2.5L 24004-2.5L-ALU
BJ24004-135KGEA 2519.8 USD InStock
BJ24004-135KG BJ24004-5L BJ24004-1L BJ24004-4X2.5L BJ24004-4X5L BJ24004-25L BJ24004-6X1L BJ240042.5LALU BJ24004-2.5L
Diethyl ether ≥99.5% (by GC) stabilized, Puriss., meets analytical specification of BP, Ph. Eur., Riedel-de Haën™
Diethyl ether
  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Beilstein Number: 1696894
RTECS Number: KI5775000
UNSPSC Code: 12191602


Formula: (CH₃CH₂)₂O
MW: 74.12 g/mol
Boiling Pt: 34-35 °C (1.013 hPa)
Melting Pt: -116 °C (1.013 hPa)
Density: 0.71 g/cm³ (20 °C)
Flash Pt: -45 °C (1.013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
https://us-legacy.vwr.com/stibo/web/std.lang.all/02/58/730258.jpg
Order Now
Specification Test Results
Refractive Index n20/D 1.3530 (lit.)
Identity A,B complying
Assay (GC) Min. 99.5 %
Boiling range 34,0 - 35,0 °C
Density (D 20/20) 0.714 - 0.716
Non-volatile matter Max. 0.002 %
Water (Karl Fischer) Max. 0.2 %
Free acid (as CH₃COOH) Max. 0.002 %
Aldehydes complying
Peroxides (as H₂O₂) Max. 0.0002 %
Acidity complying
Assay BHT (GC) 5 - 8 ppm
Residual solvents complying
Vapor Density 2.6 (vs air)

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty