Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzyl alcohol 99-100.5% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Warning
Synonyms: Phenylcarbinol

24122-4X2.5L 24122-6X1L 24122-30KG 24122-1L 24122-2.5L
BJ24122-4X2.5LCS 980.96 USD InStock
BJ24122-4X2.5L BJ24122-2.5L BJ24122-30KG BJ24122-1L BJ24122-6X1L
Benzyl alcohol 99-100.5% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
Benzyl alcohol
  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Beilstein Number: 878307
RTECS Number: DN3150000
UNSPSC Code: 12191503


Formula: C₆H₅CH₂OH
MW: 108.14 g/mol
Boiling Pt: 205.3 °C (1.013 hPa)
Melting Pt: -15.4 °C
Density: 1.045 g/cm³ (20 °C)
Flash Pt: 100.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00004599
CAS Number: 100-51-6
EINECS: 202-859-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/97/87/2299787.jpg
Order Now
Specification Test Results
Ignition Residue ≤0.005% (as SO₄)
Identity (IR) complying
Assay (GC) 99.0 - 100.5 %
Density (D 20/20) 1.043 - 1.049
Density (D 25/25) 1.042 - 1.047
Refractive index (n 20/D) 1.539 - 1.541
Non-volatile matter Max. 0.05 %
Water (Karl Fischer) Max. 0.1 %
Sulfated ash Max. 0.005 %
Free acid complying
Free acid (as C₆H₅COOH) Max. 0.02 %
Benzaldehyde (GC) Max. 0.05 %
Cyclohexylmethanol (GC) Max. 0.10 %
Sum peaks before Benzyl alcohol (GC) Max. 0.02 %
Sum peaks after Benzyl alcohol (GC) Max. 0.2 %
Related substances (GC) complying
Peroxide value Max. 5
Appearance of the solution complying
Residual solvents complying
Vapor Density 3.7 (vs air)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty