Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Acetone ≥99% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Dimethyl ketoneDimethylformaldehydeDMKPropanone2-Propanone

24201-4X5L 24201-4X2.5L 24201-6X1L 24201-4X2.5L-GL 24201-2.5L 24201-25L 24201-5L 24201-155KG 24201-20KG 24201-1L 24201-2.5L-GL 24201-10L
BJ2420142.5LGLCS 400.77 USD InStock
BJ2420142.5LGL BJ24201-4X5L BJ242012.5LGL BJ24201-4X2.5L BJ24201-20KG BJ24201-25L BJ24201-5L BJ24201-10L BJ24201-2.5L BJ24201-1L BJ24201-155KG BJ24201-6X1L
Acetone ≥99% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
Acetone
  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Beilstein Number: 635680


Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 55-57 °C (1.013 hPa)
Melting Pt: -95 °C
Density: 0.79 g/cm³ (20 °C)
Flash Pt: -17 °C
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/07/720607.jpg
Order Now
Specification Test Results
identity (IR) complying
assay (GC) Min. 99.0 %
density (D 20/4) 0.790 - 0.792
density (D 25/25) Max. 0.789
non-volatile matter Max. 20 ppm
water (Karl Fischer) Max. 0.3 %
acidity or alkalinity complying
heavy metals (as Pb) Max. 0.0001 %
related subst. (GC) complying
benzene (GC) Max. 2 Vol.ppm
methanol (GC) Max. 0.05 Vol.%
2-propanol (GC) Max. 0.05 Vol.%
reducing impurities complying
water insoluble substances complying
residual solvents complying
appearance of the solution complying
Vapor Pressure 184 mmHg ( 20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty