Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 37% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Muriatic acidHydrochloric acid solution

258148-2.5L 258148-4L 258148-6X500ML 258148-4X4L
BJ258148-4LEA 178.66 USD InStock
BJ258148-4L BJ258148-500ML BJ258148-4X4L BJ258148-2.5L
Hydrochloric acid 37% ACS, Fluka™
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 42 °C (1.013 hPa)
Melting Pt: -35 °C
Density: 1.190 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Assay 36.5 - 38.0 %
Residue on ignition (SO₄) Max. 5 ppm
Arsenic (As) Max. 0.01 ppm
Iron (Fe) Max. 0.2 ppm
Vapor Pressure 3.23 psi ( 21.1 °C)
Vapor Density 1.3 (vs air)
Heavy metals (ICP) ACS Max. 1 ppm
Ammonium (NH₄) Max. 1 ppm
Bromide (Br) Max. 50 ppm
Free chlorine (Cl) Max. 1 ppm
Sulfate (SO₄) Max. 1 ppm
Sulfite (SO₃) Max. 1 ppm
APHA Max. 10

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty