Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Dimethylacetamide ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Acetic acid dimethylamideDimethyl Acetamide

270555-4X2.5L 270555-6X1L 270555-1L 270555-4X4L
BJ270555-4X4LCS 2463.89 USD InStock
BJ270555-4X4L BJ-270555-1L BJ270555-1L BJ270555-2.5L
N,N-Dimethylacetamide ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
N,N-Dimethylacetamide
  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities

Beilstein Number: 1737614


Formula: CH₃CON(CH₃)₂
MW: 87.12 g/mol
Boiling Pt: 164-166 °C (1.013 hPa)
Melting Pt: -20 °C
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD00008686
CAS Number: 127-19-5
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/13/2298913.jpg
Order Now
Specification Test Results
IR spectrum complying
Assay (GC) Min. 99.90 %
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.03 %
Absorbance at 270 nm Max. 1.0
Absorbance 280 nm Max. 0.30
Absorbance at 290 nm Max. 0.15
Absorbance at 310 nm Max. 0.05
Absorbance at 320 nm Max. 0.03
Absorbance at 360 nm Max. 0.01
Absorbance at 400 nm Max. 0.01
Explosion Limit 1.8 %, 100 °F
Vapor Density 3 (vs air)
Explosion Limit 11.5 %, 160 °F

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty